missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Methylenebis(6-tert-butyl-p-cresol) 99.0+%, TCI America™
Supplier: TCI America M0217250G
Specifications
| 2,2′-Methylenebis(6-tert-butyl-p-cresol) | |
| 130°C | |
| C23H32O2 | |
| 250 g | |
| KGRVJHAUYBGFFP-UHFFFAOYSA-N | |
| 2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol | |
| 8398 | |
| ≥99.0% (GC) |
| 119-47-1 | |
| White-Yellow | |
| MFCD00043641 | |
| Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 3,3′-Di-tert-butyl-2,2′-dihydroxy-5,5′-dimethyldiphenylmethane, 2,2′-Methylenebis(6-tert-butyl-4-methylphenol) | |
| CC1=CC(CC2=CC(C)=CC(=C2O)C(C)(C)C)=C(O)C(=C1)C(C)(C)C | |
| 340.51 | |
| 340.51 | |
| Crystalline Powder |
Chemical Identifiers
| 119-47-1 | |
| 340.51 | |
| KGRVJHAUYBGFFP-UHFFFAOYSA-N | |
| 2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol |
| C23H32O2 | |
| MFCD00043641 | |
| 8398 | |
| CC1=CC(CC2=CC(C)=CC(=C2O)C(C)(C)C)=C(O)C(=C1)C(C)(C)C |
Safety and Handling
EINECSNumber : (4)-0100
RTECSNumber : PA3500000
TSCA : Yes