missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis[6-(benzotriazol-2-yl)-4-tert-octylphenol] 98.0+%, TCI America™

Catalog No. M1259500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1259500G 500 g
M125925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1259500G Supplier TCI America Supplier No. M1259500G
Only null left

Chemical Identifiers

CAS 103597-45-1
Molecular Formula C41H50N6O2
MDL Number MFCD00192289
Synonym Bis[3-(benzotriazol-2-yl)-2-hydroxy-5-tert-octylphenyl]methane

Specifications

Melting Point 198°C
Color Yellow
Quantity 500 g
Formula Weight 658.89
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Methylenebis[6-(benzotriazol-2-yl)-4-tert-octylphenol]
EINECSNumber (5)-5906
TSCA Yes