missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis(4-chlorophenol), TCI America™

Catalog No. M021325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M021325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M021325G Supplier TCI America Supplier No. M021325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97-23-4
Molecular Formula C13H10Cl2O2
Molecular Weight (g/mol) 269.12
MDL Number MFCD00002322
InChI Key MDNWOSOZYLHTCG-UHFFFAOYSA-N
Synonym dichlorophen, dichlorophene, dichlorofen, dichlorphen, trivex, 2,2'-methylenebis 4-chlorophenol, didroxan, didroxane, anthiphen, antiphen
PubChem CID 3037
ChEBI CHEBI:34689
IUPAC Name 4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol
SMILES OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O

Specifications

Melting Point 175°C
Quantity 25 g
UN Number 3077
Formula Weight 269.12
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Methylenebis(4-chlorophenol)
EINECSNumber (4)-0087
RTECSNumber SM0175000
TSCA Yes