missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis(4-chlorophenol), 95%

Catalog No. AAA1264036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1264036 500 g
AAA1264014 25 g
AAA1264022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1264036 Supplier Thermo Scientific Chemicals Supplier No. A1264036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-23-4 | C13H10Cl2O2 | 269.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-23-4
Molecular Formula C13H10Cl2O2
Molecular Weight (g/mol) 269.12
MDL Number MFCD00002322
InChI Key MDNWOSOZYLHTCG-UHFFFAOYSA-N
Synonym dichlorophen, dichlorophene, dichlorofen, dichlorphen, trivex, 2,2'-methylenebis 4-chlorophenol, didroxan, didroxane, anthiphen, antiphen
PubChem CID 3037
ChEBI CHEBI:34689
IUPAC Name 4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol
SMILES OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O

Specifications

Melting Point 172°C to 176°C
Quantity 500 g
UN Number UN3077
Beilstein 1884514
Merck Index 14,3071
Formula Weight 269.13
Percent Purity 95%
Chemical Name or Material 2,2'-Methylenebis(4-chlorophenol)
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-567-1
RTECSNumber SM0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only