missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyethoxy)ethanol, 98%, Spectrum™ Chemical

Catalog No. 18608151
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608151 20 L Steel Can
18608150 500 mL Amber Glass Bottle
18608149 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608151 Supplier Spectrum Chemical Mfg Cor Supplier No. M122220LTMT
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 111-77-3
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
IUPAC Name 2-(2-methoxyethoxy)ethan-1-ol
SMILES COCCOCCO

Specifications

Acidity 0.0001
CAS Max % 1
Boiling Point 193°C to 194°C (379.4°F to 381°F)
Packaging Steel Can
Quantity 20 L
Percent Purity 0.98
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-(2-Methoxyethoxy)ethanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.