missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyethyl)-1-methylpyrrolidine 98.0+%, TCI America™

Catalog No. M090825ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M090825ML 25 mL
M090810ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M090825ML Supplier TCI America Supplier No. M090825ML

Chemical Identifiers

CAS 67004-64-2
Molecular Formula C7H16NO
Molecular Weight (g/mol) 130.21
MDL Number MFCD00003174
InChI Key FYVMBPXFPFAECB-ZETCQYMHSA-O
Synonym 1-methyl-2-pyrrolidineethanol, 2-1-methylpyrrolidin-2-yl ethanol, 2-pyrrolidineethanol, 1-methyl, 2-2-hydroxyethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-ol, 2-1-methyl-2-pyrrolidine ethanol, acmc-209nxf, ksc903s0l, 1-methylpyrrolidine-2-ethanol, 1-methyl-2-pyrrolidine ethanol
PubChem CID 93363
IUPAC Name (2S)-2-(2-hydroxyethyl)-1-methylpyrrolidin-1-ium
SMILES C[NH+]1CCC[C@H]1CCO

Specifications

Boiling Point 111°C
Quantity 25 mL
Formula Weight 129.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Hydroxyethyl)-1-methylpyrrolidine