missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyethyl)-1-Methylpyrrolidine, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022904
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022904 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022904 Supplier Merck Emd Millipore Supplier No. BP526

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 67004-64-2
Density 0.951 g/mL (at 25°C (literature))
Boiling Point 110°C to 112°C (14 mmHg) (literature)
Flash Point 84°C (Closed Cup)
Molecular Formula C7H15NO
Refractive Index n20/D 1.4713 (literature)
Linear Formula C7H15NO
MDL Number MFCD00003174
Quantity 100 mg
Synonym 1-Methyl-2-pyrrolidine ethanol
Formula Weight 129.20
Grade BP
Show More Show Less