missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole 98.0+%, TCI America™

Catalog No. H0716500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0716500G 500 g
H071625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0716500G Supplier TCI America Supplier No. H0716500G
Only null left

Chemical Identifiers

CAS 3147-75-9
Molecular Formula C20H25N3O
Molecular Weight (g/mol) 323.44
MDL Number MFCD00013338
InChI Key IYAZLDLPUNDVAG-UHFFFAOYSA-N
PubChem CID 62485
IUPAC Name 2-(2H-1,2,3-benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(O)C(=C1)N1N=C2C=CC=CC2=N1

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 500 g
Formula Weight 323.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole
EINECSNumber (5)-5371
TSCA Yes