missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Furan-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine 95.0+%, TCI America™

Catalog No. F03625G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03625G Supplier TCI America Supplier No. F03625G

Chemical Identifiers

CAS 154880-05-4
Molecular Formula C11H5Cl6N3O
Molecular Weight (g/mol) 407.881
MDL Number MFCD00671535
InChI Key PNDRGJCVJPHPOZ-UHFFFAOYSA-N
Synonym 2,4-Bis(trichloromethyl)-6-[2-(furan-2-yl)vinyl]-1,3,5-triazine
PubChem CID 53405093
IUPAC Name 2-[2-(furan-2-yl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
SMILES C1=COC(=C1)C=CC2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 142°C
Color Yellow
Quantity 5 g
Formula Weight 407.88
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-[2-(Furan-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine