missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluorophenyl)pyrrolidine, 95%

Catalog No. AAH61699MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61699MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH61699MD Supplier Thermo Scientific Chemicals Supplier No. H61699MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72216-04-7 | C10H12FN | 165.211 g/mol

2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.

Chemical Identifiers

CAS 72216-04-7
Molecular Formula C10H12FN
Molecular Weight (g/mol) 165.211
MDL Number MFCD02663474
InChI Key NOJCVVKIBLHAGW-UHFFFAOYSA-N
Synonym 2-2-fluorophenyl pyrrolidine, 2-2-fluorophenyl-pyrrolidine, pyrrolidine,2-2-fluorophenyl, rs-2-2-fluoro-phenyl-pyrrolidine
PubChem CID 3613306
IUPAC Name 2-(2-fluorophenyl)pyrrolidine
SMILES C1CC(NC1)C2=CC=CC=C2F

Specifications

Color Colorless
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 165.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(2-Fluorophenyl)pyrrolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only