missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethyl acetate, 99%

Catalog No. AC257420025
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC257420025 2.5 L Glass bottle
AC257420010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC257420025 Supplier Thermo Scientific Chemicals Supplier No. 257420025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-15-2 | C8H16O4 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00041928
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
Synonym carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
PubChem CID 8165
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O

Specifications

Acidity 0.2 mg KOH/g max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -25.0°C
Color Beige to White
Density 1.0100g/mL
Boiling Point 210.0°C to 220.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4206 to 1.4226
Linear Formula CH3C(O)OCH2CH2OCH2CH2OC2H5
Quantity 2.5 L
Residue after Evaporation 0.2% max.
Specific Gravity 1.01
Merck Index 15, 3134
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, most oils
Viscosity 2.8 mPa.s (20°C)
Formula Weight 176.2
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2-(2-Ethoxyethoxy)ethyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 203-940-1

RUO – Research Use Only