missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dipyridyl N,N'-dioxide, 98%

Catalog No. AC319610050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC319610050 5 g Glass bottle
AC319610010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC319610050 Supplier Thermo Scientific Chemicals Supplier No. 319610050

CAS: 7275-43-6 | C10H8N2O2 | 188.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7275-43-6
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00065169
InChI Key FERMVCULDZOVOJ-KTKRTIGZSA-N
Synonym 2,2'-bipyridine 1,1'-dioxide, 2,2'-dipyridyl n,n'-dioxide, 2,2'-bipyridine n,n'-dioxide, 2,2'-dipyridyl-n,n'-dioxide
PubChem CID 6005843
SMILES [O-]N1C=CC=C\C1=C1/C=CC=C[N+]1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 296°C to 298°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
IUPAC Name (Z)-1-oxo-1H,1'H-1λ⁵-[2,2'-bipyridinyliden]-1-ylium-1'-olate
Formula Weight 188.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 2'-Dipyridyl N, N'-dioxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only