missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylglycine, 98%

Catalog No. AAB2265022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2265022 100 g
AAB2265006 5 g
AAB2265014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2265022 Supplier Thermo Scientific Chemicals Supplier No. B2265022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3060-50-2 | C14H13NO2 | 227.26 g/mol

2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.

Solubility
It is sparingly soluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3060-50-2
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00008048
InChI Key YBONNYNNFBAKLI-UHFFFAOYSA-N
Synonym 2,2-diphenylglycine, alpha,alpha-diphenylglycine, alpha-aminodiphenylacetic acid, glycine, 2,2-diphenyl, alpha-amino-alpha-phenylbenzeneacetic acid, .alpha.-aminodiphenylacetic acid, .alpha.,.alpha.-diphenylglycine, benzeneacetic acid, .alpha.-amino-.alpha.-phenyl, aminodiphenylacetic acid, phenytoin impurity c
PubChem CID 18289
IUPAC Name 2-amino-2,2-diphenylacetic acid
SMILES NC(C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼246°C (decomposition)
Solubility Information It is sparingly soluble in water.
Quantity 100 g
Formula Weight 227.26
Percent Purity 98%
Chemical Name or Material 2,2-Diphenylglycine
EINECSNumber 221-299-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only