missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dinitrodibenzyl 98.0+%, TCI America™

Catalog No. D26025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26025G 5 g
D260225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26025G Supplier TCI America Supplier No. D26025G

Chemical Identifiers

CAS 16968-19-7
Molecular Formula C14H12N2O4
Molecular Weight (g/mol) 272.26
MDL Number MFCD00024296
InChI Key YBOZRPPSBVIHGJ-UHFFFAOYSA-N
PubChem CID 28168
IUPAC Name 1-nitro-2-[2-(2-nitrophenyl)ethyl]benzene
SMILES C1=CC=C(C(=C1)CCC2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 5 g
Synonym 1,2-Bis(2-nitrophenyl)ethane, 2,2′C-Dinitrobibenzyl
Formula Weight 272.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dinitrodibenzyl
RTECSNumber DT4391000
TSCA No