missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylsuccinic Acid 98.0+%, TCI America™

Catalog No. D21845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21845G 5 g
D218425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21845G Supplier TCI America Supplier No. D21845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 597-43-3
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00004196
InChI Key GOHPTLYPQCTZSE-UHFFFAOYSA-N
Synonym 2,2-dimethylsuccinic acid, 2,2-dimethyl-succinic acid, unii-mx6tvr2107, 2,2-dimethyl-butanedioic acid, succinic acid, 2,2-dimethyl, butanedioic acid, 2,2-dimethyl, 2,2-dimethylbutanedioate, 2,2-dimethyl-butanedioicacid, 2,2,-dimethylsuccinate, 2,2-dimethyl succinate
PubChem CID 11701
ChEBI CHEBI:86537
IUPAC Name 2,2-dimethylbutanedioic acid
SMILES CC(C)(CC(O)=O)C(O)=O

Specifications

Melting Point 141°C
Color White
Quantity 5 g
Formula Weight 146.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethylsuccinic Acid