missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethylhexanoic Acid 98.0+%, TCI America™
$123.41 - $451.87
Chemical Identifiers
| CAS | 813-72-9 |
|---|---|
| Molecular Formula | C8H16O2 |
| Molecular Weight (g/mol) | 144.21 |
| MDL Number | MFCD00965097 |
| InChI Key | YTTWDTVYXAEAJA-UHFFFAOYSA-N |
| PubChem CID | 163217 |
| IUPAC Name | 2,2-dimethylhexanoic acid |
| SMILES | CCCCC(C)(C)C(O)=O |
Chemical Identifiers
| 813-72-9 | |
| 144.21 | |
| YTTWDTVYXAEAJA-UHFFFAOYSA-N | |
| 2,2-dimethylhexanoic acid |
| C8H16O2 | |
| MFCD00965097 | |
| 163217 | |
| CCCCC(C)(C)C(O)=O |
Specifications
| 813-72-9 | |
| 122°C | |
| MFCD00965097 | |
| 2,2-dimethyl-hexanoic acid, hexanoic acid,2,2-dimethyl, 2,2-dimethylhexanoicacid, dimethylhexanoic acid, dimethyl-hexanoic acid, acmc-209plb, 2,2-dimethyl hexanoic acid, #, hexanoic acid, 2,2-dimethyl, yttwdtvyxaeaja-uhfffaoysa | |
| CCCCC(C)(C)C(O)=O | |
| 144.21 | |
| 144.21 | |
| Liquid |
| Colorless | |
| C8H16O2 | |
| 3265 | |
| YTTWDTVYXAEAJA-UHFFFAOYSA-N | |
| 2,2-dimethylhexanoic acid | |
| 163217 | |
| ≥98.0% (GC,T) | |
| 2,2-Dimethylhexanoic Acid |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : No