missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylhexanoic Acid 98.0+%, TCI America™

Catalog No. D28091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28091G 1 g
D28095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28091G Supplier TCI America Supplier No. D28091G

Chemical Identifiers

CAS 813-72-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00965097
InChI Key YTTWDTVYXAEAJA-UHFFFAOYSA-N
PubChem CID 163217
IUPAC Name 2,2-dimethylhexanoic acid
SMILES CCCCC(C)(C)C(O)=O

Specifications

Color Colorless
Boiling Point 122°C
Quantity 1 g
UN Number 3265
Synonym 2,2-dimethyl-hexanoic acid, hexanoic acid,2,2-dimethyl, 2,2-dimethylhexanoicacid, dimethylhexanoic acid, dimethyl-hexanoic acid, acmc-209plb, 2,2-dimethyl hexanoic acid, #, hexanoic acid, 2,2-dimethyl, yttwdtvyxaeaja-uhfffaoysa
Formula Weight 144.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylhexanoic Acid
EINECSNumber (2)-0608
TSCA No