accessibility menu, dialog, popup

UserName

H3CH3CCH3OHO

CAS RN 813-72-9

IUPAC Name: 2,2-dimethylhexanoic acid
Synonyms: 2,2-dimethylhexanoic acid
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: YTTWDTVYXAEAJA-UHFFFAOYSA-N
SMILES: CCCCC(C)(C)C(O)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)