missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-4-pentenoic acid, 97%

Catalog No. AC428090010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC428090010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428090010 Supplier Thermo Scientific Chemicals Supplier No. 428090010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16386-93-9 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16386-93-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00012304
InChI Key BGUAPYRHJPWVEM-UHFFFAOYSA-N
Synonym 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g
PubChem CID 140065
IUPAC Name 2,2-dimethylpent-4-enoic acid
SMILES CC(C)(CC=C)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9330g/mL
Boiling Point 104.0°C to 108.0°C (20.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH2=CHCH2C(CH3)2COOH
Refractive Index 1.4310 to 1.4340 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.933
Formula Weight 128.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 2-Dimethyl-4-pentenoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only