missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoropropionic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH2603003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2603003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2603003 Supplier Thermo Scientific Chemicals Supplier No. H2603003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 373-96-6 | Molecular Formula: C3H4F2O2 | Molecular Weight: 110.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 373-96-6
Molecular Formula C3H4F2O2
Molecular Weight (g/mol) 110.06
MDL Number MFCD03093764
InChI Key PMWGIVRHUIAIII-UHFFFAOYSA-N
Synonym 2,2-difluoropropionic acid, 2,2-difluoropropionicacid, 2,2-difluoro-propionic acid, propanoic acid, 2,2-difluoro, acmc-1ctsu, 2,2-difluorpropionic acid
PubChem CID 2782825
IUPAC Name 2,2-difluoropropanoic acid
SMILES CC(F)(F)C(O)=O

Specifications

Melting Point 36°C to 44°C
Boiling Point 142°C
Flash Point 66°C (150°F)
Linear Formula CH3CF2COOH
Quantity 1 g
UN Number UN3261
Formula Weight 110.06
Percent Purity 97%
Chemical Name or Material 2,2-Difluoropropionic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only