missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-2-(fluorosulfonyl)acetic Acid 95.0+%, TCI America™

Catalog No. F05005G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05005G Supplier TCI America Supplier No. F05005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1717-59-5
Molecular Formula C2HF3O4S
Molecular Weight (g/mol) 178.081
MDL Number MFCD01320807
InChI Key VYDQUABHDFWIIX-UHFFFAOYSA-N
Synonym 2,2-difluoro-2-fluorosulfonyl acetic acid, 2-fluorosulfonyl difluoroacetic acid, 2-fluorosulphonyl difluoroacetic acid, difluoro fluorosulfonyl acetic acid, fluorosulfonyldifluoroacetic acid, 2,2-difluoro-2-fluorosulfonyl acetate, 2,2-difluoro-2-fluorosulfonyl-acetic acid, acetic acid, difluoro fluorosulfonyl, fluorosulfonyl difluoroacetic acid, 2,2-difluoro-2-fluoro sulfonyl acetic acid
PubChem CID 2737516
IUPAC Name 2,2-difluoro-2-fluorosulfonylacetic acid
SMILES C(=O)(C(F)(F)S(=O)(=O)F)O

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 g
UN Number 3265
Formula Weight 178.08
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 2,2-Difluoro-2-(fluorosulfonyl)acetic Acid
TSCA No
Recommended Storage Refrigerator