missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-1,3-benzodioxole 98.0+%, TCI America™

Catalog No. D40515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40515G 5 g
D405125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40515G Supplier TCI America Supplier No. D40515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1583-59-1
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00236217
InChI Key DGCOGZQDAXUUBY-UHFFFAOYSA-N
Synonym 2,2-difluorobenzo d 1,3 dioxole, 2,2-difluorobenzodioxole, 2,2-difluoro-2h-1,3-benzodioxole, 1,2-difluoromethylenedioxy benzene, 2,2-difluoro-benzodioxole, 2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 2,2-difluoro, pubchem17570, acmc-1bus1, ksc491c9d
PubChem CID 74103
IUPAC Name 2,2-difluoro-2H-1,3-benzodioxole
SMILES FC1(F)OC2=CC=CC=C2O1

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 158.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole