missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichlorocyclopropyl Phenyl Sulfide 97.0+%, TCI America™

Numéro de catalogue. D26601G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D26601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. D26601G Fournisseur TCI America Code fournisseur D26601G
Il en reste null

Identifiants chimiques

CAS 63289-85-0
Molecular Formula C9H8Cl2S
Molecular Weight (g/mol) 219.12
MDL Number MFCD00800592
InChI Key DIHSMJVOQUSOTO-QMMMGPOBSA-N
PubChem CID 2733183
IUPAC Name {[(1S)-2,2-dichlorocyclopropyl]sulfanyl}benzene
SMILES ClC1(Cl)C[C@@H]1SC1=CC=CC=C1

Spécifications

Quantity 1 g
Formula Weight 219.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,2-Dichlorocyclopropyl Phenyl Sulfide