missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-propylacetic acid, 98+%

Catalog No. AAA1296230
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1296230 250 g
AAA1296209 10 g
AAA1296218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1296230 Supplier Thermo Scientific Chemicals Supplier No. A1296230

CAS: 99-66-1 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-66-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00002672
InChI Key NIJJYAXOARWZEE-UHFFFAOYSA-N
Synonym valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
PubChem CID 3121
ChEBI CHEBI:39867
IUPAC Name 2-propylpentanoic acid
SMILES CCCC(CCC)C(O)=O

Specifications

Density 0.905
Boiling Point 219°C to 220°C
Flash Point 126°C (258°F)
Odor Weak, Characteristic
Refractive Index 1.425
Quantity 250 g
UN Number UN3265
Beilstein 1750447
Merck Index 14,9913
Formula Weight 144.21
Percent Purity ≥98%
Chemical Name or Material 2,2-Di-n-propylacetic acid
Show More Show Less
Hazard Category H302-H315-H318-H360Df
Hazard Statement GHS H Statement
H360-H314-H318-H302
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 202-777-3
RTECSNumber YV7875000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only