missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chlorophenyl)-4,5-diphenylimidazole 98.0+%, TCI America™

Numéro de catalogue. p-7125352
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C22405G 5 g
C224025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C22405G Fournisseur TCI America Code fournisseur C22405G
Il en reste null

Identifiants chimiques

CAS 1707-67-1
Molecular Formula C21H15ClN2
Molecular Weight (g/mol) 330.82
MDL Number MFCD00030328
InChI Key NSWNXQGJAPQOID-UHFFFAOYSA-N
PubChem CID 74356
IUPAC Name 2-(2-chlorophenyl)-4,5-diphenyl-1H-imidazole
SMILES ClC1=CC=CC=C1C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1

Spécifications

Melting Point 197°C
Color Yellow
Quantity 5 g
Formula Weight 330.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Chlorophenyl)-4,5-diphenylimidazole