missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bithiophene, 98%

Catalog No. AAA1233514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1233514 25 g
AAA1233503 1 g
AAA1233506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1233514 Supplier Thermo Scientific Chemicals Supplier No. A1233514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 492-97-7 | C8H6S2 | 166.256 g/mol

2,2'-Bithiophene is used as a precursor in the preparation of 5,5'-bis(trimethylstannyl)-2,2'-bithiophene , which is used for the synthesis of small molecules and polymer semiconductors for organic field-effect transistor (OFETs), organic light-emitting diode (OLED), Plastic Light Emitting Diode (PLED) and Organic Photovoltaic (OPV) applications. It is associated with aryl iodides and involved in rhodium catalyzed arylation of heteroarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Bithiophene is used as a precursor in the preparation of 5,5′-bis(trimethylstannyl)-2,2′-bithiophene , which is used for the synthesis of small molecules and polymer semiconductors for organic field-effect transistor (OFETs), organic light-emitting diode (OLED), Plastic Light Emitting Diode (PLED) and Organic Photovoltaic (OPV) applications. It is associated with aryl iodides and involved in rhodium catalyzed arylation of heteroarenes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 492-97-7
Molecular Formula C8H6S2
Molecular Weight (g/mol) 166.256
MDL Number MFCD00005414
InChI Key OHZAHWOAMVVGEL-UHFFFAOYSA-N
Synonym 2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene
PubChem CID 68120
ChEBI CHEBI:36821
IUPAC Name 2-thiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC=CS2

Specifications

Melting Point 30°C to 33°C
Boiling Point 260°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 3039
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 166.27
Percent Purity 98%
Chemical Name or Material 2,2'-Bithiophene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-767-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only