missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-oxocyclohexyl)propane 95.0+%, TCI America™

Catalog No. p-7119199
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16725G 5 g
B167225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16725G Supplier TCI America Supplier No. B16725G

Chemical Identifiers

CAS 7418-16-8
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00191618
InChI Key HAWVCXABNZBPED-UHFFFAOYSA-N
PubChem CID 81914
IUPAC Name 4-[2-(4-oxocyclohexyl)propan-2-yl]cyclohexan-1-one
SMILES CC(C)(C1CCC(=O)CC1)C2CCC(=O)CC2

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 5 g
Synonym 4,4′C-Isopropylidenebis(cyclohexanone)
Formula Weight 236.36
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-oxocyclohexyl)propane
EINECSNumber (4)-1726
TSCA No