missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomers) 93.0+%, TCI America™

Catalog No. B092825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B092825G 25 g
B0928500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B092825G Supplier TCI America Supplier No. B092825G
Only null left

Chemical Identifiers

CAS 80-04-6
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.387
MDL Number MFCD00019334
InChI Key CDBAMNGURPMUTG-UHFFFAOYSA-N
PubChem CID 94932
IUPAC Name 4-[2-(4-hydroxycyclohexyl)propan-2-yl]cyclohexan-1-ol
SMILES CC(C)(C1CCC(CC1)O)C2CCC(CC2)O

Specifications

Color White
Quantity 25 g
Synonym p,p′C-Dihydroxy-2,2-dicyclohexylpropane, Perhydrobisphenol A
Formula Weight 240.39
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomers)
EINECSNumber (4)-0201
TSCA Yes