missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-hydroxy-3-methylphenyl)propane 98.0+%, TCI America™

Catalog No. B1567500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1567500G 500 g
B156725G 25 g
B1567100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1567500G Supplier TCI America Supplier No. B1567500G
Only null left

Chemical Identifiers

CAS 79-97-0
Molecular Formula C17H20O2
Molecular Weight (g/mol) 256.345
MDL Number MFCD00002232
InChI Key YMTYZTXUZLQUSF-UHFFFAOYSA-N
PubChem CID 6620
ChEBI CHEBI:34321
IUPAC Name 4-[2-(4-hydroxy-3-methylphenyl)propan-2-yl]-2-methylphenol
SMILES CC1=C(C=CC(=C1)C(C)(C)C2=CC(=C(C=C2)O)C)O

Specifications

Melting Point 139°C
Color White
Quantity 500 g
Synonym 4,4′-Isopropylidenedi-o-cresol
Formula Weight 256.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-hydroxy-3-methylphenyl)propane
EINECSNumber (4)-1696
RTECSNumber GP1750000
TSCA Yes