missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(4-acetoxyphenyl)propane 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 92°C |
| Color | White |
| Boiling Point | 215°C |
| Quantity | 25 g |
| Formula Weight | 312.37 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2-Bis(4-acetoxyphenyl)propane |
Sécurité et manipulation
| EINECSNumber | (4)-1661 |
| TSCA | Yes |