missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridine, 99+%

Catalog No. AAA1578214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1578214 25 g
AAA1578222 100 g
AAA1578236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1578214 Supplier Thermo Scientific Chemicals Supplier No. A1578214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol

Reagent for iron determinations.2,2'-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for iron determinations.2,2′-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

Solubility
Soluble in organic solvents and slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition.

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
PubChem CID 1474
ChEBI CHEBI:30351
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

Melting Point 70°C
Boiling Point 272°C to 273°C
Flash Point 121°C (249°F)
Quantity 25 g
UN Number UN2811
Beilstein 113089
Merck Index 14,3347
Solubility Information Soluble in organic solvents and slightly soluble in water.
IUPAC Name 2,2'-bipyridine
Formula Weight 156.19
Percent Purity ≥99%
Chemical Name or Material 2,2'-Bipyridine
Show More Show Less
Hazard Category H301-H312-H500
Hazard Statement GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-674-4
RTECSNumber DW1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only