missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bipyrazine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 188°C |
| Color | Yellow |
| Quantity | 500 mg |
| Synonym | 2,2′-Bipyrazinyl |
| Formula Weight | 158.16 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2′-Bipyrazine |
Safety and Handling
| TSCA | No |