missing translation for 'onlineSavingsMsg'
Learn More

2,2', Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis, MP Biomedicals™

Catalog No. ICN19502305
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19502305 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19502305 Supplier MP Biomedicals Inc Supplier No. 0219502305

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30931-67-0
Molecular Formula C18H24N6O6S4
Molecular Weight (g/mol) 548.67
MDL Number MFCD00010404,MFCD00010404
InChI Key OHDRQQURAXLVGJ-AXMZSLBLSA-N
Synonym ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
PubChem CID 91884754
IUPAC Name diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
SMILES [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O

Specifications

Color Green
Quantity 5 g
Formula Weight 548.7
Physical Form Powder
Chemical Name or Material 2, 2' Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis