missing translation for 'onlineSavingsMsg'
Learn More

2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene 98.0+%, TCI America™

Catalog No. T27591G
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
T27591G 1 g
1 options

Catalog No. T27591G

Supplier: TCI America T27591G

Only null left

Chemical Identifiers

CAS 27192-91-2
Molecular Formula C26H12Br4
Molecular Weight (g/mol) 643.998
InChI Key ZPFZLDRAZNLKJI-UHFFFAOYSA-N
PubChem CID 13806537
IUPAC Name 2,7-dibromo-9-(2,7-dibromofluoren-9-ylidene)fluorene
SMILES C1=CC2=C(C=C1Br)C(=C3C4=C(C=CC(=C4)Br)C5=C3C=C(C=C5)Br)C6=C2C=CC(=C6)Br

Specifications

Melting Point 455°C
Quantity 1 g
Formula Weight 644.00
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′,7,7′-Tetrabromo-9,9′-bifluorenylidene