missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl 98.0+%, TCI America™
Supplier: TCI America T18071G
Specifications
| 2,2′,6,6′-Tetra-tert-butyl-4,4′-dihydroxybiphenyl | |
| 186°C | |
| C28H42O2 | |
| 1 g | |
| GSOYMOAPJZYXTB-UHFFFAOYSA-N | |
| 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol | |
| 67185 | |
| ≥98.0% (GC) |
| 128-38-1 | |
| Yellow | |
| MFCD00069443 | |
| 2,2′C,6,6′C-Tetra-tert-butyl-[1,1′C-biphenyl]-4,4′C-diol | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C | |
| 410.64 | |
| 410.64 | |
| Crystalline Powder |
Chemical Identifiers
| 128-38-1 | |
| 410.64 | |
| GSOYMOAPJZYXTB-UHFFFAOYSA-N | |
| 67185 | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C |
| C28H42O2 | |
| MFCD00069443 | |
| 2,2′C,6,6′C-Tetra-tert-butyl-[1,1′C-biphenyl]-4,4′C-diol | |
| 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol |
Safety and Handling
TSCA : No