accessibility menu, dialog, popup

UserName

CAS RN 128-38-1

IUPAC Name: 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol
Synonyms: 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol
Molecular Weight (g/mol): 410.64
Molecular Formula: C28H42O2
InChi Key: GSOYMOAPJZYXTB-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results