missing translation for 'onlineSavingsMsg'
Learn More

2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl 98.0+%, TCI America™

Catalog No. p-7136970
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1807200MG 200 mg
T18071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1807200MG Supplier TCI America Supplier No. T1807200MG
Only null left

Chemical Identifiers

CAS 128-38-1
Molecular Formula C28H42O2
Molecular Weight (g/mol) 410.64
MDL Number MFCD00069443
InChI Key GSOYMOAPJZYXTB-UHFFFAOYSA-N
Synonym 2,2′C,6,6′C-Tetra-tert-butyl-[1,1′C-biphenyl]-4,4′C-diol
PubChem CID 67185
IUPAC Name 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 186°C
Color Yellow
Quantity 200 mg
Formula Weight 410.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′,6,6′-Tetra-tert-butyl-4,4′-dihydroxybiphenyl