missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine, 96%

Catalog No. AC215632500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC215632500 250 mg
AC215630010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC215632500 Supplier Thermo Scientific Chemicals Supplier No. 215632500

CAS: 1148-79-4 | C15H11N3 | 233.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1148-79-4
Molecular Formula C15H11N3
Molecular Weight (g/mol) 233.27
MDL Number MFCD00006213
InChI Key DRGAZIDRYFYHIJ-UHFFFAOYSA-N
Synonym 2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine
PubChem CID 70848
ChEBI CHEBI:245199
IUPAC Name 2,6-dipyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1

Specifications

Melting Point 89.0°C to 91.0°C
Boiling Point 370.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Quantity 250 mg
Formula Weight 233.28
Percent Purity 96%
Chemical Name or Material 2,2':6',2''-Terpyridine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF exposed or concerned:
EINECSNumber 214-559-5

RUO – Research Use Only