missing translation for 'onlineSavingsMsg'
Learn More

acide benzoïque 2-(2,5-diméthyl-1H-pyrrol-1-yl), 97%, Thermo Scientific™

Numéro de catalogue. CC39901CB
Modifier l'affichage
Click to view available options
Quantité:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC39901CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC39901CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC39901CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 92028-57-4
Formule moléculaire C13H13NO2
Poids moléculaire (g/mol) 215.25
Numéro MDL MFCD01125264
Clé InChI ZLYUUANOICYAAL-UHFFFAOYSA-N
Synonyme 2-2,5-dimethyl-1h-pyrrol-1-yl benzoic acid, 2-2,5-dimethylpyrrol-1-yl benzoic acid, 2-2,5-dimethyl-1-pyrrolyl benzoic acid, 2-2,5-dimethyl-pyrrol-1-yl-benzoic acid, 2-2,5-dimethylpyrrolyl benzoic acid, cbmicro_048042, benzoic acid,2-2,5-dimethyl-1h-pyrrol-1-yl
PubChem CID 292956
Nom de l’IUPAC 2-(2,5-diméthylypyrrol-1-yl)acide benzoïque
SOURIRES CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O

Spécifications

Nom ou substance chimique 2-(2, 5-Dimethyl-1H-pyrrol-1-yl)benzoic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 250 mg
Poids de formule 215.25
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.