missing translation for 'onlineSavingsMsg'
Learn More

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide, 99%

Catalog No. AAA1829406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1829406 5 g
AAA1829403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1829406 Supplier Thermo Scientific Chemicals Supplier No. A1829406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19805-75-5 | C9H16N2O | 168.24 g/mol

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide may be used in the synthesis of 2,2,5,5-tetramethyl-3-pyrrolidinecarboxamide and 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy free radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide may be used in the synthesis of 2,2,5,5-tetramethyl-3-pyrrolidinecarboxamide and 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy free radical.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 19805-75-5
Molecular Formula C9H16N2O
Molecular Weight (g/mol) 168.24
MDL Number MFCD00041846
InChI Key ACFYUJLIWIDSFM-UHFFFAOYSA-N
Synonym 2,2,5,5-tetramethyl-3-pyrroline-3-carboxamide, 2,2,5,5-tetramethyl-2,5-dihydro-1h-pyrrole-3-carboxamide, 1h-pyrrole-3-carboxamide, 2,5-dihydro-2,2,5,5-tetramethyl, 2,5-dihydro-2,2,5,5-tetramethyl-1h-pyrrole-3-carboxamide, 3-aminocarbonyl-2,2,5,5-tetramethy-3-pyrroline, maybridge3_000440, acmc-1c3ma, 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrroline, 3-aminocarbonyl-2,2,5,5-tetramethyl-3-pyrroline
PubChem CID 88255
IUPAC Name 2,2,5,5-tetramethyl-1H-pyrrole-3-carboxamide
SMILES CC1(C=C(C(N1)(C)C)C(=O)N)C

Specifications

Melting Point 179°C to 183°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 168.24
Percent Purity 99%
Chemical Name or Material 2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 243-331-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only