missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane 99.0+%, TCI America™

Catalog No. T0715250ML
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0715250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0715250ML Supplier TCI America Supplier No. T0715250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Color Colorless
Quantity 250 mL
Physical Form Clear Liquid at 20°C
UN Number 1262
Boiling Point 99°C
Formula Weight 114.23
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2,2,4-Trimethylpentane [for Spectrophotometry]
EINECSNumber (2)-0008
RTECSNumber SA3320000
TSCA Yes