missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, Exceeds A.C.S. Specifications, HPLC Grade, 99%7, Spectrum™ Chemical

Catalog No. 18608153
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608153 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18608153 Supplier Spectrum Chemical Mfg Cor Supplier No. HP8924LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 540-84-1
Molecular Weight (g/mol) 114.23

Specifications

CAS Max % 1
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 3 ppm
Water 0.0002
Boiling Point 99.238°C
Percent Purity 0.997
Grade ACS/HPLC
Chemical Name or Material 2,2,4-Trimethylpentane

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.