missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Triméthylpentane, 99+%, réactif ACS
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | 2, 2, 4-Trimethylpentane |
| Point de fusion | -107.0°C |
| Nom Remarque | For analysis ACS |
| CAS % minimum | 99.0 |
| CAS % maximal | 100.0 |
| Densité | 0.6900g/mL |
| Quantité | 1 L |
| Point d’éclair | −12°C |
| Emballage | Bouteille en verre |
| Forme physique | Liquide |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: May be fatal if swallowed and enters airways. Very toxic to aquatic life with long lasting effects. May cause drowsiness or dizziness. Causes skin irritation. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Usage réservé à la recherche