missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%

Catalog No. AC422000050
Change view
Click to view available options
Quantity:
5 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422000050 5 g Glass bottle
AC422000010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422000050 Supplier Thermo Scientific Chemicals Supplier No. 422000050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6846-50-0 | C16H30O4 | 286.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6846-50-0
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.4
MDL Number MFCD00059267
InChI Key OMVSWZDEEGIJJI-UHFFFAOYSA-N
Synonym 2,2,4-trimethyl-1,3-pentanediol diisobutyrate, kodaflex txib, 2,2,4-trimethylpentane-1,3-diyl bis 2-methylpropanoate, txib, propanoic acid, 2-methyl-, 2,2-dimethyl-1-1-methylethyl-1,3-propanediyl ester, 2,2,4-trimethylpentanediol-1,3-diisobutyrate, isobutyric acid, 1-isopropyl-2,2-dimethyltrimethylene ester, 1-isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-trimethyl-1,3-pentanediol ester, dsstox_cid_7635
PubChem CID 23284
IUPAC Name [2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate
SMILES CC(C)C(C(C)(C)COC(=O)C(C)C)OC(=O)C(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless
Density 0.9400g/mL
Boiling Point 280.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 5 g
Specific Gravity 0.94
Merck Index 15, 9399
Solubility Information Solubility in water: insoluble
Formula Weight 286.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
Show More Show Less
EINECSNumber 229-934-9
RTECSNumber SA1420000
TSCA TSCA

RUO – Research Use Only