missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,6,6-Pentaméthylheptane

Numéro de catalogue. AC462050050
Modifier l'affichage
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC462050050 5 mL
AC462050250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC462050050 Fournisseur Thermo Scientific Chemicals Code fournisseur 462050050
Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

CAS : 13475-82-6 | C12H26 | 170,34 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Identifiants chimiques

CAS 13475-82-6
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key VKPSKYDESGTTFR-UHFFFAOYSA-N
IUPAC Name 2,2,4,6,6-pentaméthéthylheptane
SMILES CC(CC(C)(C)C)CC(C)(C)C

Spécifications

Density 0.7463 g/mL
Boiling Point 170.0°C
Flash Point 37°C
Linear Formula [CH3C(CH3)2CH2]2CHCH3
Quantity 5 mL
Specific Gravity 0.7463
Formula Weight 170.33
Chemical Name or Material 2,2,4,6,6-Pentamethylheptane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Flammable liquid and vapor.

May be fatal if swallowed and enters airways.

May cause long lasting harmful effects to aquatic life.

Repeated exposure may cause skin dryness or cracking.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Do NOT induce vomiting.

Store in a well-ventilated place.

Keep container tightly closed.

EINECSNumber 236-757-0

RUO – Usage réservé à la recherche