missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,8,8-Heptamethylnonane, 98%

Catalog No. AC156252500
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156252500 250 mL Glass bottle
AC156250500 50 mL Glass bottle
AC156250010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156252500 Supplier Thermo Scientific Chemicals Supplier No. 156252500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4390-04-9 | C16H34 | 226.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4390-04-9
Molecular Formula C16H34
Molecular Weight (g/mol) 226.44
MDL Number MFCD00008856
InChI Key VCLJODPNBNEBKW-UHFFFAOYNA-N
Synonym isocetane, nonane, 2,2,4,4,6,8,8-heptamethyl, cyprane, pubchem16042, acmc-2097fb, dsstox_cid_30668, dsstox_gsid_52101, ksc492g9j, 2,4,4,6,8,8-heptamethylnonane, nonane,2,4,4,6,8,8-heptamethyl
PubChem CID 20414
IUPAC Name 2,2,4,4,6,8,8-heptamethylnonane
SMILES CC(CC(C)(C)C)CC(C)(C)CC(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.7900g/mL
Boiling Point 240.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400
Linear Formula (CH3)3CCH2CH(CH3)CH2C(CH3)2CH2C(CH3)3
Quantity 250 mL
Specific Gravity 0.79
Solubility Information Solubility in water: insoluble
Formula Weight 226.44
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,8,8-Heptamethylnonane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
I
EINECSNumber 224-506-8

RUO – Research Use Only