missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,4,4,4-Hexafluoro-1-butanol 94.0+%, TCI America™

Catalog No. H064925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H064925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H064925G Supplier TCI America Supplier No. H064925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 382-31-0
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.065
MDL Number MFCD00042309
InChI Key LVFXLZRISXUAIL-UHFFFAOYSA-N
Synonym 2,2,3,4,4,4-hexafluoro-1-butanol, 1h,1h,3h-hexafluorobutanol, 2,2,3,4,4,4-hexafluorobutanol, 2,2,3,4,4,4-hexafluorobutanol-1, 2,2,3,4,4,4-hexafluoro-butan-1-ol, 1-butanol, 2,2,3,4,4,4-hexafluoro, 1h,1h,3h-perfluorobutan-1-ol, acmc-209iys, cf3cfhcf2ch2oh, 2,2,3,4,4,4-hexafluorobutyl alcohol
PubChem CID 533990
IUPAC Name 2,2,3,4,4,4-hexafluorobutan-1-ol
SMILES C(C(C(C(F)(F)F)F)(F)F)O

Specifications

Color Yellow
Boiling Point 113°C
Quantity 25 g
Formula Weight 182.07
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,4,4,4-Hexafluoro-1-butanol