missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D

Catalog No. AAA14489MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14489MD 250 mg
AAA1448903 1 g
AAA1448906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA14489MD Supplier Thermo Scientific Chemicals Supplier No. A14489MD

CAS: 24493-21-8 | C6H13NaO2Si | 172.267 g/mol

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

Solubility
Soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 24493-21-8
Molecular Formula C6H13NaO2Si
Molecular Weight (g/mol) 172.267
MDL Number MFCD00002762
InChI Key OIIWPAYIXDCDNL-HGFPCDIYSA-M
Synonym sodium 3-trimethylsilyl 2,2,3,3-2h4 propionate, 3-trimethylsilyl propionic acid-d4 sodium salt, 2,2,3,3-d4-3-trimethylsilyl propionic acid sodium salt, sodium 3-trimethylsilyl 2 h? propanoate, sodium 3-trimethylsilyl 2h4 propanoate, tsp-d4, propanoic-2,2,3,3-d4 acid, 3-trimethylsilyl-, sodium salt, sodium 3-trimethylsilyl propionate-2,2,3,3-d4, 3-trimethylsilyl propionic-2,2,3,3-d4 acid sodium salt, sodium-3-trimethylsilyl-2,2,3,3-tetradeuteriopropionate
PubChem CID 23688921
IUPAC Name sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate
SMILES C[Si](C)(C)CCC(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Linear Formula (CH3)3SiCD2CD2CO2Na
Quantity 250 mg
Beilstein 4046239
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 172.27
Assay ≥98 atom %D
Chemical Name or Material 2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-286-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only