missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol 98.0+%, TCI America™

Catalog No. O011425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O011425G 25 g
O0114100G 100 g
O0114500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O011425G Supplier TCI America Supplier No. O011425G

Chemical Identifiers

CAS 355-80-6
Molecular Formula C5H4F8O
Molecular Weight (g/mol) 232.07
MDL Number MFCD00039631
InChI Key JUGSKHLZINSXPQ-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-octafluoro-1-pentanol, 1h,1h,5h-octafluoro-1-pentanol, 1-pentanol, 2,2,3,3,4,4,5,5-octafluoro, 1,1,5-trihydrooctafluoropentyl alcohol, 2,2,3,3,4,4,5,5-octafluoropentanol, 1,1,5-trihydrooctafluoropentan-1-ol, unii-d05k8ejx15, 1h,1h,5h-octafluoropentanol, 2,2,3,3,4,4,5,5-octafluoropentyl alcohol, alpha,alpha,omega-trihydroperfluoropentanol
PubChem CID 9641
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)F

Specifications

Color Colorless
Boiling Point 140°C
Quantity 25 g
Formula Weight 232.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
RTECSNumber SA7650000
TSCA Yes