missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptyloxirane 98.0+%, TCI America™

Numéro de catalogue. T33655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T33655G 5 g
T336525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T33655G Fournisseur TCI America Code fournisseur T33655G
Il en reste null

Identifiants chimiques

CAS 38565-52-5
Molecular Formula C9H5F13O
Molecular Weight (g/mol) 376.12
MDL Number MFCD00042356
InChI Key KGYUZRBIQCDOCN-UHFFFAOYNA-N
Synonym 3-Perfluorohexyl-1,2-epoxypropane
PubChem CID 170073
IUPAC Name 2-(2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptyl)oxirane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1

Spécifications

Color Colorless
Boiling Point 154°C
Quantity 5 g
Formula Weight 376.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptyloxirane